Find methods for your needs
AN114: Determination of Trace Anions in High-Purity Waters Using Direct Injection and Two-Step Isocratic Ion Chromatography
Description
The determination of trace anionic contamination is important to power generation and to the electronics industries to minimize corrosion. Here µg/L concentrations of anions are determined in a 1000-µL injection, in pull mode, of a morpholine solution on IonPac AS14 column. This application is optimized for a fast run time (10 min) using a two-step isocratic separation at two concentration levels of boric acid and sodium hydroxide eluents. AN114 is also suitable for trace anions in pure water. AU191 describes a similar technique to analyze samples containing boric acid and lithium.| Market: | Water Analysis | 
| Keywords: | Formate, morpholine, fluoride, isocratic, power plant, IonPac AS14, Nitrate, boric acid eluent, Sodium hydroxide eluent, Chloride, Oxalate, phosphate, Acetate, Sulfate, Glycolate, Carbonate, AN 114, AU 191, AU191 | 
| Matrix: | Morpholine | 
| Author: | Ed Kaiser and Jeff Rohrer | 
For Research Use Only. Not for use in diagnostic procedures.