Find methods for your needs
AN66: Determination of Neomycin B and Impurities Using HPAE-IPAD
Description
In this application note, we combine the CarboPac PA1, an eluent generator with CR-ATC, and disposable Au working electrodes to demonstrate an improved HPAE-PAD technology for Neomycin B purity analysis and its determination in Neosporin topical ointment, a complex over-the-counter pharmaceutical formulation.| Market: | BioPharma |
| Keywords: | aminoglycoside, carbohydrate, CarboPac PA1, Electrochemical detection, Eluent Generation, nebramine, Pharmaceutical, neamine, ointment, HPAE-PAD, TN 20, TN 70671, TN20, TN70671, AN 66 |
| Matrix: | Ointment |
| Author: | Valoran Hanko and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.