Find methods for your needs
TN126: Determination of Organic Acids in Beer Samples Using a High-Pressure Ion Chromatography System
Description
This Technical Note presents the advantages of the 4 μm particle-size IonPac AS11-HC-4μm column combined with the Dionex ICS-5000+ HPIC system for optimal separation of organic acids and inorganic anions in beer samples using electrolytically generated hydroxide eluent with and without an organic modifier. An ICS-6000 can be used for this application.| Market: | Food and Beverage |
| Keywords: | Formate, Fumarate, Malate, Pyruvate, Quinate, Organic acids, Citrate, HPIC, ICS-5000+ HPIC system, IonPac AS11-HC-4μm column, Aconitate, fluoride, Succinate, cis-Aconitate, Isocitrate, trans-Aconitate, Beer, RFIC, Chloride, Lactate, Oxalate, phosphate, Sulfate, Carbonate, ICS-6000 |
| Matrix: | Degassed beer samples |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.