Find methods for your needs
AN150: Determination of Amino Acids in Cell Cultures and Fermentation Broths.
Description
This application note describes the use of AAA-Direct to analyze common amino acids in the presence of simple sugars, sugar alcohols, alcohols, and glycols in yeast and bacterial fermentation broths. In this application note, the AminoPac PA10 anion-exchange column is used to separate amino acid and carbohydrate ingredients in cell culture and fermentation broth media. High carbohydrate concentrations can hamper the determination of some amino acids due to coelution. Also in this application note, disposable gold (Au) electrodes are evaluated for use in amino acid determination.| Market: | BioPharma |
| Keywords: | AAA-Direct, Amino acids, AminoPac PA10, Carbohydates, Cell Cultures, Disposable gold electrodes, Fermentation Broths, Integrated pulsed amperometric detection (IPAD), IPAD, sugars |
| Matrix: | Cell culture, Fermentation broth media |
| Author: | Valoran Hanko and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.