Find methods for your needs
AN270: Determination of Hydroxymethylfurfural (HMF) in Honey and Biomass
Description
This work describes a high-performance anion-exchange chromatography with pulsed amperometric detection (HPAE-PAD)-based method for the determination of HMF in samples ranging from food to treated biomass. A Dionex ICS-3000 system with a CarboPac PA1 column, electrolytically generated hydroxide eluent, and electrochemical detection with disposable Au-on-polytetrafluoroethylene working electrodes are used. The Dionex CarboPac PA1 is a high-capacity, rugged column suitable for determining mono- and disaccharides, and has high resolution for HMF in a wide variety of matrices.| Market: | Food and Beverage |
| Keywords: | 5-hydroxymethylfurfural, biofuel, biofuels, Carbohydrates, CarboPac PA1, corn stover, disposable electrodes, EG, Eluent Generation, food spoilage, HMF, hydroxymethylfurfural, syrup, Honey, PAD, HPAE-PAD, TN 20, TN 70671, TN20, TN70671, AN 270 |
| Matrix: | Biomass, Corn stover, Honey |
| Author: | Lipika Basumallick and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.