Find methods for your needs
Improved monitoring of monoclonal antibody heterogeneity by cation-exchange chromatography using a Thermo Scientific ProPac WCX-10 column
Description
The Thermo Scientific UltiMate 3000 titanium system is applied for analyzing a humanized IgG1 MAb for C-terminal lysine variants. The separation was performed on a Thermo Scientific ProPac WCX-10 column, with UV detection at 280 nm.| Market: | BioPharma |
| Keywords: | Biocompatible, HPLC, Lysine, MAb, Monoclonal antibody, ProPac WCX-10, Protein, UV detection, Variants |
| Matrix: | Humanized IgG (mAb) |
For Research Use Only. Not for use in diagnostic procedures.