Find methods for your needs
Fast analysis of water-soluble vitamins on a Thermo Scientific Acclaim RSLC PolarAdvantage II (PA2) column
Description
The Thermo Scientific Dionex UltiMate 3000 RSLC is applied for the analysis of water soluble vitamins from soft drink. The separation was performed in less than 5 minutes on a Thermo Scientific Acclaim RSLC PolarAdvantage II (PA2) 2.2 µm column with UV detection.| Market: | Food and Beverage |
| Keywords: | Acclaim PolarAdvantage II (PA2), Ascorbic acid, Citrate, Folic acid, Niacin, Niacinamide, Pantothenic acid, Pyridoxine, Riboflavin, Soft drink, Thiamine, Water-soluble vitamins |
| Matrix: | Soft drink |
For Research Use Only. Not for use in diagnostic procedures.