Find methods for your needs
Rapid analysis of soft drinks using a Thermo Scientific Acclaim Mixed-Mode WAX-1 column
Description
The Thermo Scientific Acclaim Mixed-Mode WAX-1 column is used for the rapid analysis of soft drink ingredients, with UV detection at 210 nm. Due to the columns multi-functional chemistry, this assay can be performed in just 3 minutes.| Market: | Food and Beverage |
| Keywords: | Acclaim Mixed-Mode WAX-1, Acesulfame, Aspartame, benzoate, Caffeine, Citrate, HPLC, Rapid analysis, Soft drink, Sorbate, Uv detection |
| Matrix: | Diet Cola |
For Research Use Only. Not for use in diagnostic procedures.