Find methods for your needs
AN72209: Determination of Trace Concentrations of Oxyhalides and Bromide in Municipal and Bottled Waters
Description
To demonstrate that oxyhalides and bromide can be successfully determined at concentrations required for regulatory standards and guidelines using a Thermo Scientific™ Dionex™ ion chromatography system equipped with a Thermo Scientific™ Dionex™ IonPac™ AS23-4μm column,Thermo Scientific™ Dionex™ CRD 300 Carbonate Removal Device, and a Thermo Scientific™ Dionex™ AERS™ 500 Carbonate Anion Electrolytically Regenerated Suppressor.| Market: | Environmental; Food and Beverage |
| Keywords: | Bromate, Chlorate, Chlorite, Disinfection Byproducts, HPIC, suppressed conductivity detection, RFIC System, Drinking water analysis, US EPA 300.1 Part B, Dionex IonPac AS23-4μm column, Dionex AERS 500 Carbonate Suppressor, Dionex CRD 300, carbonate removal device |
| Matrix: | Water |
| Author: | Jingli Hu and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.