Find methods for your needs
Separation of oxidized and deamidated impurities of Infliximab by reverse phase chromatography
Description
The Thermo Scientific Dionex UltiMate 3000 Bio-LC system is applied for the Reverse phase analysis of Infliximab. The separation was performed on a Thermo Scientific MabPac RP column with UV detection at 214 nm. All oxidized and deamidated impurities are well separated.| Market: | BioPharma |
| Keywords: | HPLC, Monoclonal antibody, UV detection, MAbPac RP, Infliximab |
| Matrix: | N/A |
| Author: | Krishnarao Dara |
| Affiliation: | Thermo Fisher Scientific, CCS CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.