Find methods for your needs
AN72210: Fast Determination of Biofuel Sugars by HPAE-PAD
Description
This work updates the column used in AN1161 with a shorter 4 × 150 mm format column. The combination of the smaller resin particle size of the Dionex CarboPac SA10-4μm column and the shorter column used here results in separation of eight common sugars in less than six minutes. This is significant time savings as compared to the eight-minute runtime achieved in AN1089. The shorter run time allows for faster sample turnaround times and reduced eluent consumption, thereby improving the overall process economics. Results for method linearity, accuracy, and robustness are presented here.Market: | Energy |
Keywords: | Electrochemical detection, Monosaccharides, Fermentable sugars, HPAE-PAD, Carbopac SA10-4μm, Lignocellulosic biomass, Biomass hydrolysis, Biomass-to-biofuel conversion, TN 20, TN 70671, TN20, TN70671, AN 72210 |
Matrix: | DI water |
Author: | Sachin Patil and Jeffrey Rohrer (Only first and corresponding authors shown; for complete author list refer to the attached application document text) |
Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA 94085 |
For Research Use Only. Not for use in diagnostic procedures.