Find methods for your needs
AN1161: Improved Method for Determination of Biofuel Sugars by HPAE-PAD
Description
The current work updates the system used for the carbohydrate analysis in AN1089. The new system combines flexibility and ease-of-use with high sensitivity and selectivity, bringing a higher level of convenience and cost effectiveness to simple sugar analysis. The method proposed here separates eight common carbohydrate sugars in less than eight minutes. The shorter run time results in better process economics. Using this method, carbohydrates present in 10 individual biofuel samples were quantified.| Market: | Petrochem, Oil and Gas |
| Keywords: | Electrochemical detection, Fermentable sugars, Carbopac SA10-4μm, Dionex Integrion HPIC System, Lignocellulosic biomass, Biomass hydrolysis, Biomass-to-biofuel conversion, TN 20, TN 70671, TN20, TN70671, AN 1161 |
| Matrix: | Water |
| Author: | Sachin Patil and Jeffrey Rohrer (Only first and corresponding author shown; for complete author list refer to the attached application document text) |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA 94085 |