Find methods for your needs
Separation of Acidic and Basic charge variants of Denosumab by Ion Exchange chromatography using Salt Gradient Method
Description
The Thermo Scientific Dionex UltiMate 3000 LC Bio-LC system is applied for the Ion Exchange analysis of Denosumab. The separation was performed on a Thermo Scientific MabPac SCX-10 column with a UV detection at 220 nm. With a run time of 65 minutes the method allows easy separation of charge variants of Denosumab.| Market: | BioPharma | 
| Keywords: | HPLC, MAbPac SCX-10 Column, Denosumab, PDA detection, Charge Variants, Salt Gradient | 
| Matrix: | N/A | 
| Author: | Manish Singh | 
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India | 
For Research Use Only. Not for use in diagnostic procedures.