Find methods for your needs
Analysis of Protein Therapeutics and Excipients using 2D Chromatography with Combined UV and Charged Aerosol Detection Be the first to rate this application
analysis protein therapeutics excipients using 2d chromatography with combined uv charged aerosol detection
Description
Therapeutic proteins (antibodies and vaccines) vary considerably and unwanted aggregation is a major path to degradation. Stabilization of protein formulations can be enhanced through the addition of specific excipients such as amino acids, ions, surfactants and sugars. A 2D “heart cut” technique allows the separation of the protein component of a commercial therapeutic formulation and analyzed using a DAD detector and polar sugar and amino acid excipients to be separated on the second dimension using ion-pairing RP conditions and detected via the charged aerosol detector.
Market: BioPharma
Matrix: Water
Author: Bruce Bailey
Affiliation: Thermo Scientific
Uploaded on 12/3/2015.
For Research Use Only. Not for use in diagnostic procedures.