Find methods for your needs
AN1144: Ion Chromatography Assay for Lithium in Lithium Hydroxide
Description
The IC-based method described in this application note uses a Thermo Scientific Dionex IonPac CS16 cation-exchange column, an electrolytically generated MSA eluent, and suppressed conductivity detection to determine lithium in lithium hydroxide. The method proposed in this application note was validated following the guidelines outlined in USP General Chapter <1225>, Validation of Compendial Procedures to meet the requirements for lithium and calcium quantification prescribed in the lithium hydroxide USP monograph. An ICS-6000 can be used for this application.| Market: | Pharma |
| Keywords: | ICS-5000+, IonPac CS16, Pharmaceutical, USP monograph, RFIC, CERS 500, AN 1144, ICH, ICS-6000 |
| Matrix: | DI water |
| Author: | Sachin Patil and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA, USA |
For Research Use Only. Not for use in diagnostic procedures.