Find methods for your needs
AU152: An Improved Gradient Method for the AAA-Direct™ Separation of Amino Acids and Carbohydrates in Complex Sample Matrices
Description
In this Application Update, we combine into a single method individual improvements made to the standard AAA-Direct gradient program. These individual improvements are now in the AAA-Direct manual. This update shows that such combined changes to the AAA-Direct method improve the performance of our previously published method. This new method successfully determines amino acids in yeast extract-peptone-dextrose yeast culture medium (YPD Broth), and Dulbecco’s modified Eagle’s (with F-12), M199, L-15 (Leibovitz), and McCoy’s 5A media for mammalian cell culture.| Market: | BioPharma; Clinical Research; Proteomics |
| Keywords: | PAD, AminoPac PA10, AU152, Citrulline, Hydroxylysine, Ornithine, Taurine, Inositol |
| Matrix: | DI water |
| Author: | Valoran Hanko and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.