Description
In bioformulations containing a protein and a carbohydrates, carbohydrates are used to assist in the stabilization of the proteins in solution, and are used in different amounts, depending on the protein and the product requirements. Sorbitol, a sugar alcohol is transferred to and analysed by HILIC and CAD after it is eluted from the reverse-phase column. Both sorbitol and the protein are analysed in this 2D-HPLC method.| Market: | BioPharma |
| Keywords: | Bovine Serum Albumin (BSA), carbohydrate, Protein, Sorbitol |
| Matrix: | Water |
| Author: | Marc Plante |
| Affiliation: | Thermo Scientific |
For Research Use Only. Not for use in diagnostic procedures.