Find methods for your needs
CAN114: Quantification of Trace and Major Anions in Water by Ion Chromatography in a High-Throughput Laboratory
Description
Laboratories split their methods because good resolution for the major inorganic anion peaks and a low LOD for bromate determination is a difficult compromise to achieve. This application is a 2 channels method for the determination of fluoride, chloride, chlorite, chlorate, bromide, bromate, nitrite, nitrate, sulfate, and phosphate in different types of water samples without compromise between the sample volume injected in the chromatographic system and a low limit of quantification suitable for trace bromate determinations. An ICS-6000 can be used for this application.| Market: | Water Analysis |
| Keywords: | Bromate, Chlorate, Chlorite, fluoride, anions, Nitrate, RFIC, ion chromatography, Nitrite, Chloride, phosphate, Sulfate, Anion-exchange, Suppressed Conductivity, trace analysis, Bromide, Dionex ICS-5000+ dual system, Dionex IonPac AS19, Dionex IonPac AS9-HC, Trace bromate, ICS-6000 |
| Matrix: | Water |
| Author: | Sébastien N. Ronkart |
| Affiliation: | Société wallonne des eaux. (www.swde.be) |
For Research Use Only. Not for use in diagnostic procedures.