Find methods for your needs
TN30: Monosaccharide and Oligosaccharide Analysis of Glycoproteins Electrotransferred onto Polyvinylidene Fluoride (PVDF) Membranes
Description
This older TN shows how HPAE-PAD carbohydrate analysis can be performed from a PVDF blot. The TN shows monosaccharide, oligosaccharide, and sialic acid analyses. This record reports the monosaccharide analysis method. For updated monosaccharide, sialic acid, and oligosaccharide analysis methods please see records for TN40, AU180, AU181, and AN1050.| Market: | BioPharma |
| Keywords: | carbohydrate, CarboPac PA1, Electrochemical detection, glycoprotein, oligosaccharide, sialic acid, PAD, monosaccharide, HPAE-PAD, CarboPac PA100, TN71, TN 20, TN 70671, TN20, TN70671, TN 30 |
| Matrix: | Glycoprotein |
| Author: | Michael Weitzhandler |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.