Find methods for your needs
CAN118: Determination of Trace Sodium in Diesel and Biodiesel Fuel
Description
Sodium determination in diesel fuel using inductively coupled plasma-optical emission spectrometry (ICP-OES) is possible; limits of detection are ~0.1 ppm. The goal of this study was to develop and test an alternate approach using IC for the determination of cations in diesel fuel. The approach focused on the use of commercially available modules and existing standard chromatographic conditions. The challenge was to design an automated, cost-effective, fast, and sensitive method that can provide 10–100 times greater sensitivity than ICP-OES method and does not require manual extraction steps.| Market: | Petrochem, Oil and Gas |
| Keywords: | in-line sample preparation, IonPac CS12A Column, Reagent Free Ion Chromatography, biodiesel, RFIC, Trace Sodium, suppressed conductivity detection, diesel fuel, TCC-ULP1 Trace Cation Concentrator Column, EGC MSA Eluent Generator Cartridge, Fuels |
| Matrix: | Biodiesel fuel, Diesel |
| Author: | Joachim Trick, Cornelia Wanner |
| Affiliation: | Daimler AG, Stuttgart, Germany |
For Research Use Only. Not for use in diagnostic procedures.