Description
This method using the IonPac AS22-Fast column provides an equivalent method to the EPA 300.0 (Part A) and 300.1 (Part A) approved methods while providing a 5 min run time that reduces cycle time, lowering the overall cost, saving time, and increasing the sample throughput. This method uses the ICS-1100 Integrated IC System but can be performed easily on any other Dionex IC system, including Reagent-Free IC (RFIC) with eluent regeneration and eluent generation IC systems.| Market: | Water Analysis |
| Keywords: | Bromide, Chloride, Drinking Water, EPA 300.0, EPA 300.1, Fluoride, Ion chromatography, IonPac AS22-Fast column, Nitrate, Nitrite, Phosphate, RFIC, Sulfate, Suppressed conductivity detection |
| Matrix: | Water |
For Research Use Only. Not for use in diagnostic procedures.