Find methods for your needs
AB125: Determination of Trace Levels of Inorganic Anions in High-Purity Water Using Capillary Ion Chromatography
Description
Converting IC methods from analytical to capillary scale provides user with advantages. For example, the system can be left on and ready to run samples, the capillary eluent generation cartridge (EGC) lasts much longer. This application demonstrates the measurement of trace levels of ionic impurities in water using capillary ion chromatography with the Dionex IonPac AS15 column. This is critical for the identification and prevention of corrosive conditions in many power plant components.| Market: | Water Analysis |
| Keywords: | Anion, Bromide, Capillary ion chromatography system, Chloride, Fluoride, High-purity water, IonPac AS15 column, Nitrate, Nitrite, Phosphate, RFIC, Sulfate, Suppressed conductivity detection |
| Matrix: | Water |
For Research Use Only. Not for use in diagnostic procedures.