Find methods for your needs
TN112: Determination of Trace Anions in Ultrapure Water Using Capillary Ion Chromatography: LARGE LOOP INJECTION
Description
In technical note TN112, trace anion determinations in ultrapure water by capillary IC are discussed. Both large volume direct injection using a large loop and using a concentrator column are discussed. Additionally installation tips are discussed.| Market: | Water Analysis |
| Keywords: | Capillary, fluoride, anions, Bromide, cap IC, ion chromatography, IC, acetate, capillary IC, chloride, formate, IonPac AS19 column, nitrate, nitrite, phosphate, semiconductor, sulfate, trace analysis, ultra pure water, trace |
| Matrix: | Ultra pure water |
| Author: | Christison, Pang, Madden, Schibler, Hoefler, Lopez |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.