Find methods for your needs
AN1091: Determination of Uronic Acids and Wood Sugars in Wood-Based Hydrolysates: Comprehensive Method
Description
Wood, grass, and other lignocellulosic biomass are emerging as the preferred sources for biofuel and biochemical production. Compositional analysis of plant derived material is crucial for maximizing the yield of biofuel production. A single HPAE-PAD method, which uses the Dionex CarboPac PA20 column, can be used for the simultaneous determination of mono- and disaccharides and uronic acids.| Market: | Other |
| Keywords: | arabinose, biofuel, CarboPac PA20, Electrochemical detection, fucose, Glucose, rhamnose, xylose, Mannose, Sucrose, Cellobiose, Galacturonic Acid, glucuronic acid, fructose, galactose, HPAE-PAD, Lignocellulosic biomass, TN21, TN71, TN 71, TN 20, TN 70671, TN20, TN70671 |
| Matrix: | Wood |
| Author: | Lipika Basumallick and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.