Find methods for your needs
AN1136: Determination of Perchlorate in Drinking Water using a Microbore Reagent Free Ion Chromatography System (Part A)
Description
This Application Note expands the scope of the EPA Method 314.0 to the 2-mm column format. It evaluates and describes the linear range and initial demonstration of capability. Initial demonstration of capability includes determination of method accuracy, precision, detection limit (MDL), and matrix conductivity threshold (MCT). This document also describes perchlorate recovery in field samples. Overall, the results presented here meet or exceed the performance requirements specified in U.S. EPA Method 314.0. (This part has an one-click workflow for ICS2100 and AS-AP Autosampler)| Market: | Environmental |
| Keywords: | RFIC, IonPac AS16, EPA 314.0, MCT Verification, PHG, CCL, AN1136 |
| Matrix: | Water |
| Author: | Sachin Patil and Jeffrey Rohrer |
| Affiliation: | Thermo Scientific, Sunnyvale, CA, USA |
For Research Use Only. Not for use in diagnostic procedures.