Find methods for your needs
AN130: Identification of a Hydroxylysine-ContainingPeptide Using AAA-Direct
Description
This application note shows that AAA-Direct can be used to determine peptides with lysine hydroxylation. All common amino acids, including hydroxylysine (Hyl), were separated by AAA-Direct, and the presence or absence of Hyl in the fractions confirmed peak identities. The techniques described in this application note show several ways that peptides can be evaluated for identity and purity, how peptides can be desalted, and how much desalting is needed for successful AAADirect amino acid analysis.| Market: | BioPharma |
| Keywords: | Acclaim C18, AminoPac PA10, AN 130, CD4 fragment, ProPac WCX-10 |
| Matrix: | Water |
For Research Use Only. Not for use in diagnostic procedures.