Find methods for your needs
Quick analysis of arginine and methylated arginines by LC-MS
Description
The Thermo Scientific HPLC system is applied for the analysis of arginine and methylated arginines. The separation is performed on a Thermo Scientific Hypercarb column, with MS detection.| Market: | BioPharma |
| Keywords: | Arginine, Dimethyl arginine, HPLC, Hypercarb, L-arginine, LC-MS, Methyl-L-arginine, MS detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.