Find methods for your needs
AN1050: Evaluating Protein Glycosylation in Limited-Quantity Samples by HPAE-PAD, Monosaccharide Method
Description
HPAE-PAD methods provide a cost-effective option for investigating glycans without the derivatization required by many other methods. Electrochemical detection determines the glycans directly without the potential loss of sialylation or linkage rearrangement that may occur during analysis with other methods. HPAE-PAD can be used for determining monosaccharides, oligosaccharides, sialic acids, and other carbohydrates. This record shows a monosaccharide method.| Market: | BioPharma |
| Keywords: | CarboPac PA20, fucose, Glucose, glycoprotein, Mannose, human serum transferrin, PAD, prostate specific antigen, PSA, galactosamine, monosaccharide, galactose, glucosamine, HPAE-PAD, TN71, TN 71, TN 20, TN 70671, TN20, TN70671, A1F, charged glycans, Neutral glycans, G0F, G1F, G2bF, Man3, Man6, G0, G0bF, G1bF, Man9, A2F, G2F |
| Matrix: | Glycoprotein |
| Author: | Deanna Hurum, Lipika Basumallick, and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.