Description
Paromomycin is an aminoglycoside antibiotic produced by Streptomyces rimosus var. paromomycinus. This Application Note demonstrates the use of an electrolytically generated potassium hydroxide eluent combined with the CarboPac PA1 anion-exchange column and IPAD using a disposable AAA Au working electrode for the determination of paromomycin in a bulk pharmaceutical formulation.| Market: | Pharma |
| Keywords: | CarboPac PA1, Electrochemical detection, Eluent Generation, IPAD, Pharmaceutical, Humatin, HPAE-PAD, Drug, TN 20, TN 70671, TN20, TN70671, AN 186, AN186 |
| Matrix: | Drug |
| Author: | Brian De Borba, Valoran P. Hanko, and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.