Find methods for your needs
AN2967: Implementation of a Walk-Up High-Pressure Capillary Ion Chromatograph for the Fast Separation of Pharmaceutical Relevant Inorganic Cations (Cholesterol tablet)
Description
The counter ion impacts the API drug’s efficacy, solubility, and toxicity, and therefore counter ion determinations are important to pharmaceutical companies during all stages of product development and as part of product quality monitoring. In this example, the calcium counter ion is determined in a water extract of a pharmaceutical drug tablet designed to treat high cholesterol serum levels. Here fast calcium determinations were demonstrated on IonPac CS12A-8µm capillary column using a walk-up high pressure capillary IC system.| Market: | Pharma |
| Keywords: | capIC, capillary IC, counter cations, counter ions, fast IC, IonPac CS12A column, IonPac CS12A-8um column, cholesterol drug |
| Matrix: | Water extract of a pharmaceutical drug tablet developed to treat high cholesterol blood levels |
| Author: | T. Christison, J.E. Madden, F. Pang, and K. Divan |
For Research Use Only. Not for use in diagnostic procedures.