Find methods for your needs
UHPLC Method Development for Simultanous Determination of Antihypertensive Combination Agents
Description
The current trend in antihypertensiion therapy is the medication with several antihypertension agents in a single pill. Here eluent switching capabilities of the Thermo Scientific Vanquish Binary pump H are applied to develop a UHPLC method for simultanous determination of antihypertensive combination agents on a Thermo Scientific Accucore Vanquish C18+ column by scouting different eluent combinations.| Market: | Pharma |
| Keywords: | API, Generic Method, UHPLC Instrumentation, Automated method scouting, Vanquish UHPLC System, Diuretics |
| Matrix: | N/A |
| Author: | Susanne Fabel |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.