Find methods for your needs
AN106: Ion Chromatography in the Pharmaceutical Industry (Cations)
Description
Ion Chromatography is a simple and accurate technique for the analysis and quantification of charged species. This application note describes the use of ion exchange chromatography and ion exclusion chromatography in the pharmaceutical industry for the analysis of ions in ingredients and final products. The topic of cleaning validation is also discussed as it applies to charged species. Here shows the separation of six standard cations by cation exchange chromatography using the IonPac CS12A column with suppressed conductivity detection. This method can also be run with a RFIC system.| Market: | Pharma |
| Keywords: | Ammonium, Calcium, Cations, IC, IonPac CS12A column, Lithium, Magnesium, methanesulfonic acid, Potassium, Sodium, Suppressed conductivity, RFIC |
| Matrix: | Drug |
For Research Use Only. Not for use in diagnostic procedures.