Find methods for your needs
AN117 - Quantification of Carbohydrates and Glycols in Pharmaceuticals - Glycols Method Be the first to rate this application
an117 quantification carbohydrates glycols pharmaceuticals glycols method
Description
This application note describes the use of three different anion-exchange columns with amperometric detection to analyze common simple sugars, sugar alcohols, and glycols in pharmaceutical formulations. The method in this entry shows how to determine glycols using ion-exclusion chromatography on Dionex ICE-AS1 column.
Market: Pharma
Keywords: cold/flu formulation, Glycerol, glycol, ion-exclusion chromatography, IonPac ICE-AS1, propylene glycol, sorbitol, platinum working electrode
Matrix: Pharmaceutical formulations
Author: Valoran Hanko and Jeff Rohrer
Affiliation: Thermo Fisher Scientific
Uploaded on 4/21/2015.
For Research Use Only. Not for use in diagnostic procedures.