Find methods for your needs
AN117 - Quantification of Carbohydrates and Glycols in Pharmaceuticals - Glycols Method
Description
This application note describes the use of three different anion-exchange columns with amperometric detection to analyze common simple sugars, sugar alcohols, and glycols in pharmaceutical formulations. The method in this entry shows how to determine glycols using ion-exclusion chromatography on Dionex ICE-AS1 column.| Market: | Pharma |
| Keywords: | cold/flu formulation, Glycerol, glycol, ion-exclusion chromatography, IonPac ICE-AS1, propylene glycol, sorbitol, platinum working electrode |
| Matrix: | Pharmaceutical formulations |
| Author: | Valoran Hanko and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.