Description
Methylamine or monomethylamine (MMA) is widely used in different stages of the preparation of various drug substances and may be retained in the drug substance as process-related or degradation impurities. The work shown here uses a RFIC system with suppressed conductivity detection to determine MMA in drug products. This method uses a Thermo Scientific™ Dionex™ IonPac™ CS19 analytical column with methanesulfonic acid (MSA) eluent produced by an eluent generator. The method separates MMA from other cations typically present in drug products.Market: | Pharma |
Keywords: | Dionex IonPac CS19 Column, Impurities, Methylamine, MMA, Monomethylamine, Pharmaceutical, Reagent-Free Ion Chromatography, RFIC |
Matrix: | Drug |
For Research Use Only. Not for use in diagnostic procedures.