Find methods for your needs
Fast analysis of organic acids in orange juice on a Thermo Scientific Acclaim Organic Acid (OA) column
Description
The Thermo Scientific HPLC system is applied for the analysis of organic acids in orange juice. The separation is performed on a Thermo Scientific Acclaim Organic Acid (OA) column, with UV detection at 210 nm. The samples were prepared using Thermo Scientific Dionex OnGuard II P SPE cartridges.Market: | Food and Beverage |
Keywords: | OnGuard II P, Solid phase extraction, SPE, Acclaim Organic Acid, Ascorbic acid, Citric acid, Fumaric acid, HPLC, Malic acid, Orange juice, Organic acids, Vitamin C |
Matrix: | Orange juice |
For Research Use Only. Not for use in diagnostic procedures.