Rate
Ask the Expert
Suggest an Application
Add to Favorites
Create
Parts List
Parts List
Tell a
Friend
Friend
Print
PDF
Find methods for your needs
AN253: HPAE-PAD Determination of Infant Formula Sialic Acid
Description
Sialic acids in infant formulas are accurately determined by HPAE-PAD using the Dionex CarboPac PA20 column following acid hydrolysis and maltodextrin removal using one of two sample-preparation methods. HPAE-PAD provides reliable determination of sialic acids in acid hydrolyzed infant formula samples without sample derivatization. This method may be used to quantify sialic acids in formulas that have been enriched with sialic acids.Market: | Food and Beverage |
Keywords: | AN 253, CarboPac PA20, Human Milk, Maltodextrins, N-acetylneuraminic acid, NANA, Neu5Ac, Neu5Gc, N-glycolylneuraminic Acid, NGNA, TN 20, TN 70671, TN20, TN70671 |
Matrix: | Water |
Author: | Deanna Hurum and Jeffrey Rohrer |
Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.