Find methods for your needs
AN243: Determination of Common Anions and Organic Acids Using Ion Chromatography-Mass Spectrometry
Description
This application note demonstrates the use of the IC-MS method for the determination of five common anions and three organic acids at parts per billion levels. With the use of very specific MS SIM detection, ppb level quantification can be performed with greater confidence on real world samples, such as bottled drinking water. The quantification results from MS were without prior sample preparation or employing preconcentration, which may be required with other detection methods.| Market: | Water Analysis |
| Keywords: | AN 243, Chloride, IC-MS, IonPac AS20, MSQ Plus, Phosphate, Pyruvic acid, RFIC, Sulfate, Tartaric acid, α-ketoglutaric acid, ICMS |
| Matrix: | Water |
For Research Use Only. Not for use in diagnostic procedures.