Find methods for your needs
AN1054: Determination of Ammonia in Tobacco Smoke Be the first to rate this application
ammonia tobacco smoke
Description
The application describes an IC method that achieves better resolution between ammonium and other amines for the determination of ammonia in tobacco smoke. The IC method uses a Dionex IonPac CS19 column set with electrolytically generated MSA gradient. The moderately high capacity of the Dionex IonPac CS19 column allows sample analysis without dilution. The method has high precisions (RSD <0.2% for retention time and <1% for peak area), a low detection limit (0.01 mg/L), and good recoveries (86–115%).
Market: Food and Beverage
Keywords: Ammonium, Calcium, Cigarette, Diethylamine, Dionex IonPac CS19 Column, Ethylamine, Freebasing Nicotine, Ion Chromatography, Magnesium, Methylamine, Potassium, Propylamine, RFIC, Sodium
Matrix: Food
Author: Lipika Basumallick and Jeff Rohrer
Affiliation: Thermo Fisher Scientific
Uploaded on 2/12/2015.
For Research Use Only. Not for use in diagnostic procedures.