Find methods for your needs
AN67: Determination of Plant-Derived Neutral Oligo- and Polysaccharides
Description
This application note describes how to develop an HPAE-PAD method for separating plant-derived neutral oligo- and polysaccharides. Amylopectin and fructan separations demonstrate column, eluent, and gradient choices for developing rugged HPAE-PAD methods for neutral oligo- and polysaccharides.Market: | Food and Beverage |
Keywords: | amylopectin, amylose, carbohydrate, CarboPac PA1 column, CarboPac PA100 column, Electrochemical detection, fructan, inulin, maltodextrin, oligosaccharide, polysaccharide, MALTRIN, Beer, amber beer, chicory, dahlia, Maltrin MO40, PAD, HPAE-PAD, TN71, TN 71, TN 20, TN 70671, TN20, TN70671, AN 67 |
Matrix: | Plants |
Author: | Jeffrey Rohrer |
Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.