Find methods for your needs
AN158: Determination of Trace Sodium and Transition Metals in Power Industry Samples by Ion Chromatography with Nonsuppressed Conductivity Detection
Description
This application compares suppressed to nonsuppressed conductivity detection for the determination of sub-ppb concentrations of sodium in simulated power plant matrices. Linearity, limits of detection, and recovery of sub-ppb sodium spiked into simulated samples containing high concentrations of ethanolamine are reported. In addition, this application describes the determination of transition metals by nonsuppressed conductivity detection.| Market: | Other |
| Keywords: | Nuclear Power Plants, Preconcentration, Thermo Scientific Dionex IonPac SCS-1 Column, Transition Metals, non-suppressed conductivity, Thermo Scientific Dionex IonPac SCS 1 column |
| Matrix: | Ethanolamine |
| Author: | Brian De Borba and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.