Find methods for your needs
Fast determination of additives in carbonated beverages using a Thermo Scientific Acclaim Mixed-mode WAX-1 HPLC column Be the first to rate this application
fast determination additives carbonated beverages using a thermo scientific acclaim mixedmode wax1 hplc column
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the determination of additives in carbonated beverages. The separation is performed on a Thermo Scientific Acclaim Mixed-mode WAX-1 HPLC column, with UV detection at 210 nm.
Market: Food and Beverage
Keywords: Acclaim Mixed-Mode WAX-1, Acesulfame, Additives, Aspartame, benzoate, Caffeine, Carbonated beverages, Citrate, HPLC, Saccharin, Sorbate, UltiMate 3000, UV detection
Matrix: Beverage
Uploaded on 2/22/2010.
For Research Use Only. Not for use in diagnostic procedures.