Find methods for your needs
AN197: Determination of Glucosamine in Dietary Supplements Using HPAE-PAD
Description
Glucosamine (GlcN), a naturally occurring amino sugar, is a major structural component in the biosynthesis of glycosaminoglycans. Use of GlcN as a dietary supplement in the management of osteoarthritis has attracted considerable attention. Pulsed amperometric detection (PAD) is sensitive and selective and therefore ideally suited for determination of GlcN. High-performance anion-exchange with pulsed amperometric detection (HPAE-PAD) chromatography is a sensitive, direct-detection technique capable of separating mono- and disaccharides rapidly and efficiently.| Market: | Food and Beverage |
| Keywords: | Electrochemical detection, glucosamine, TN71, AN 197, TN 20, TN 70671, TN20, TN70671 |
| Matrix: | Dietary supplements |
| Author: | Richard Kornfeld and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.