Find methods for your needs
Fast analysis of water-soluble vitamin standards on a Thermo Scientific Acclaim PolarAdvantage II (PA2) RSLC column with TFA mobile phase additive Be the first to rate this application
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the analysis of water-soluble vitamin standards. The separation is performed on a Thermo Scientific Acclaim PolarAdvantage II (PA2) RSLC column, with UV detection at 210 nm.
Market: Food and Beverage
Keywords: Acclaim PolarAdvantage II (PA2), Ascorbic acid, Folic acid, HPLC, Niacin, Niacinamide, Pantothenic acid, Pyridoxine, Riboflavin, Thiamine, UltiMate 3000 RSLC, UV detection, Water-soluble vitamins
Matrix: N/A
Uploaded on 2/19/2010.
For Research Use Only. Not for use in diagnostic procedures.