Find methods for your needs
Fast analysis of water-soluble vitamins in a lemon soft drink using a Thermo Scientific Acclaim PolarAdvantage II (PA2) RSLC column with TFA mobile phase additive Be the first to rate this application
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the analysis of water-soluble vitamins in a lemon soft drink. The separation is performed on a Thermo Scientific Acclaim PolarAdvantage II (PA2) RSLC column, with UV detection at 210 nm.
Market: Food and Beverage
Keywords: Acclaim PolarAdvantage II (PA2), Ascorbic acid, Citrate, HPLC, Niacinamide, Pantothenic acid, Pyridoxine, UltiMate 3000 RSLC, UV detection, Water-soluble vitamins
Matrix: Propel ® (PepsiCo) Lemon
Uploaded on 2/19/2010.
For Research Use Only. Not for use in diagnostic procedures.