Find methods for your needs
Improved gradient separation of basic, neutral, and acidic pharmaceuticals on a Thermo Scientific Acclaim Mixed-Mode WAX-1 HPLC column
Description
The Thermo Scientific Acclaim Mixed-Mode WAX-1 column is used for the gradient separation of basic, neutral and acidic pharmaceuticals.The novel column chemistry of the Acclaim Mixed-Mode WAX-1 column provides a multi-mode separation mechanism that includes hydrophobic, anion-exchange and cation-exclusion interactions.| Market: | Pharma |
| Keywords: | Acclaim Mixed-Mode WAX-1, Acids, Bases, HPLC, Neutrals, Pharmaceutical, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.