Find methods for your needs
Improved analysis of choline, acetylcholine, and succinylcholine using a Thermo Scientific Acclaim Trinity P1 column with charged aerosol detection
Description
The Thermo Scientific Dionex Ultimate 3000 RSLC system with quaternary LPG is applied for the analysis of anesthetic and muscle relaxants. The separation was performed in less than 12 minutes on a Thermo Scientific Dionex Acclaim Trinity P1 3 µm column with a Thermo Scientific Dionex Corona Ultra charged aerosol detector (CAD).| Market: | Pharma |
| Keywords: | Acclaim Trinity P1, anesthetic, Charged aerosol detector, choline, HPLC, muscle relaxant |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.