Find methods for your needs
Simultaneous determination of metformin and its chloride counterion using multi-mode liquid chromatography with charged aerosol detection
Description
This work demonstrates the determination of metformin (API) and its counterion (chloride) using a simple isocratic method. Thermo Scientific Acclaim Trinity P2 columns offer cation-exchange, anion-exchange and HILIC retention mechanisms on the same phase, thus they are ideal for separating both cationic and anionic species simultaneously.| Market: | Pharma |
| Keywords: | Counterion, HPLC, Metformin, Charged aerosol detection, Chloride |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.