Find methods for your needs
Simultaneous separation of pharmaceutical counterions by HPLC-CAD Be the first to rate this application
Description
The Thermo Scientific Acclaim Trinity P2 column provides desired selectivity for the separation of mono- and multi-valent anions and cations – a total of twelve ions including sodium, potassium, magnesium, calcium, chloride, bromide, nitrate, malate, citrate, sulfate, fumareate and citrate are well separated on a 50 mm long column using a gradient method within 23 minutes.
Market: Pharma
Keywords: HPLC, Charged aerosol detection, Acclaim Trinity P2, Pharmaceutical counterions
Matrix: N/A
Uploaded on 6/19/2014.
For Research Use Only. Not for use in diagnostic procedures.