Find methods for your needs
Quaternary surfactant analysis of a cosmetic formulation using HPLC-CAD
Description
This method provides an effective analysis of quaternary surfactants in a cosmetic formulation using a Thermo Scientific Dionex UltiMate 3000 HPLC system with charged aerosol detection (CAD). A Thermo Scientific Acclaim Surfactant Plus column is used, which provides improved results as this group of surfactants tends to tail on conventional silica-based reversed-phase columns. The Thermo Scientific Corona charged aerosol detector is an effective choice since the two active ingredients in this formulation are unsuited to UV detection.| Market: | Other |
| Keywords: | Charged aerosol detector, Acclaim Surfactant Plus, Hexadecyltrimethylammonium, Quaternary surfactants, Sodium, Chloride, Hexadecanoylamidopropyltrimethylammonium, Octadecanoylamidopropyltrimethylammonium |
| Matrix: | Cosmetic formulation |
For Research Use Only. Not for use in diagnostic procedures.